Tisk…

Ethyl-3-methyl-4,4,4-trifluorobutyrate 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC3288B-1G PC3288B-5G
APOSPC3288B-1GEA 2690 CZK
APOSPC3288B-1G APOSPC3288B-5G
Ethyl-3-methyl-4,4,4-trifluorobutyrate 97%
Ethyl-3-methyl-4,4,4-trifluorobutyrate
Vzorec: C₇H₁₁F₃O₂
Bod varu: 135…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00040847
CAS číslo: 6975-13-9
UN: 3272
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR