Tisk…

4-(Trifluoromethyl)pyridine-1-oxide

Dodavatel: Apollo Scientific

Synonyms: 4-(Trifluoromethyl)pyridine 1-oxide

PC32879-1G PC32879-250MG PC32879-500MG
APOSPC32879-1GEA 14720 CZK
APOSPC32879-1G APOSPC32879-250MG APOSPC32879-500MG
4-(Trifluoromethyl)pyridine-1-oxide
4-(Trifluoromethyl)pyridine-1-oxide
Vzorec: C₆H₄F₃NO
MW: 163,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16988752
CAS číslo: 22253-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR