5-Fluoro-2-(1H-pyrrol-1-yl)pyridine
Dodavatel: Apollo Scientific
PC32878-1G
PC32878-250MG
PC32878-500MG
APOSPC32878-1GEA
9690
CZK
APOSPC32878-1G
APOSPC32878-250MG
APOSPC32878-500MG
5-Fluoro-2-(1H-pyrrol-1-yl)pyridine
5-Fluoro-2-(1H-pyrrol-1-yl)pyridine
Vzorec:
C₉H₇FN₂ Storage Temperature: Ambient |
MDL Number:
MFCD19706043 CAS číslo: 1355334-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...