1-[6-(3-Fluorophenyl)pyridin-3-yl]ethanone
Dodavatel: Apollo Scientific
PC32877-1G
PC32877-100MG
PC32877-250MG
PC32877-500MG
PC32877-2.5G
APOSPC32877-1GEA
18620
CZK
APOSPC32877-1G
APOSPC32877-100MG
APOSPC32877-250MG
APOSPC32877-500MG
APOSPC32877-2.5G
1-[6-(3-Fluorophenyl)pyridin-3-yl]ethanone
1-[6-(3-Fluorophenyl)pyridin-3-yl]ethanone
Vzorec:
C₁₃H₁₀FNO Storage Temperature: Ambient |
MDL Number:
MFCD12404911 CAS číslo: 1216541-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...