Tisk…

2-Hydroxy-4-fluorobenzoyl chloride

Dodavatel: Apollo Scientific

PC32864-1G PC32864-250MG PC32864-500MG
APOSPC32864-1GEA 14520 CZK
APOSPC32864-1G APOSPC32864-250MG APOSPC32864-500MG
2-Hydroxy-4-fluorobenzoyl chloride
2-Hydroxy-4-fluorobenzoyl chloride
Vzorec: C₇H₄ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD13173890
CAS číslo: 57976-99-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR