1-(4-Fluorophenyl)pyrrolidine-3-carboxylic acid
Dodavatel: Apollo Scientific
PC32863-1G
PC32863-100MG
PC32863-250MG
PC32863-500MG
PC32863-2.5G
PC32863-5G
APOSPC32863-1GEA
22220
CZK
APOSPC32863-1G
APOSPC32863-100MG
APOSPC32863-250MG
APOSPC32863-500MG
APOSPC32863-2.5G
APOSPC32863-5G
1-(4-Fluorophenyl)pyrrolidine-3-carboxylic acid
1-(4-Fluorophenyl)pyrrolidine-3-carboxylic acid
Vzorec:
C₁₁H₁₂FNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09864924 CAS číslo: 933731-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...