Difluoro[3-(trifluoromethyl)pyridin-2-yl]acetic acid
Dodavatel: Apollo Scientific
PC32861-1G
PC32861-50MG
PC32861-250MG
PC32861-500MG
PC32861-2.5G
APOSPC32861-1GEA
18820
CZK
APOSPC32861-1G
APOSPC32861-50MG
APOSPC32861-250MG
APOSPC32861-500MG
APOSPC32861-2.5G
Difluoro[3-(trifluoromethyl)pyridin-2-yl]acetic acid
Difluoro[3-(trifluoromethyl)pyridin-2-yl]acetic acid
Vzorec:
C₈H₄F₅NO₂ MW: 241.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12963754 CAS číslo: 1215579-44-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...