1-(2,6-Difluorophenyl)cyclobutanecarboxylic acid
Dodavatel: Apollo Scientific
PC32860-1G
PC32860-250MG
PC32860-500MG
PC32860-2.5G
PC32860-5G
APOSPC32860-1GEA
7610
CZK
APOSPC32860-1G
APOSPC32860-250MG
APOSPC32860-500MG
APOSPC32860-2.5G
APOSPC32860-5G
1-(2,6-Difluorophenyl)cyclobutanecarboxylic acid
1-(2,6-Difluorophenyl)cyclobutanecarboxylic acid
Vzorec:
C₁₁H₁₀F₂O₂ MW: 212.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11037022 CAS číslo: 1215754-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...