Tisk…

N-(3,5-Difluorobenzyl)-N-methylamine

Dodavatel: Apollo Scientific

PC32854-1G PC32854-250MG PC32854-500MG
APOSPC32854-1GEA 8370 CZK
APOSPC32854-1G APOSPC32854-250MG APOSPC32854-500MG
N-(3,5-Difluorobenzyl)-N-methylamine
N-(3,5-Difluorobenzyl)-N-methylamine
Vzorec: C₈H₉F₂N
Storage Temperature: Ambient
MDL Number: MFCD11212394
CAS číslo: 90390-28-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR