Oil 10 S
Dodavatel: Apollo Scientific
|
Varování
|
PC32827-100G
PC32827-25G
APOSPC32827-100GEA
2730
CZK
APOSPC32827-100G
APOSPC32827-25G
Oil 10 S
Oil 10 S
Inert gear box oil
Vzorec:
(C₂ClF₃)n Hustota: 1,95 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS číslo:
9002-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...