2-Bromo-3',5'-bis(trifluoromethyl)acetophenone 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(3,5-Bis(trifluoromethyl)phenyl)-2-bromoethanone, 3,5-Bis(trifluoromethyl)phenacyl bromide
PC3276-25G
PC3276-5G
APOSPC3276-25GEA
5790
CZK
APOSPC3276-25G
APOSPC3276-5G
2-Bromo-3',5'-bis(trifluoromethyl)acetophenone 97%
2-Bromo-3',5'-bis(trifluoromethyl)acetophenone
Vzorec:
C₁₀H₅BrF₆O MW: 335,0405 g/mol Bod tání: 48…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792434 CAS číslo: 131805-94-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...