Tisk…

4-Fluoro-2-formylphenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Fluoro-2-formylbenzeneboronic acid, (4-Fluoro-2-formylphenyl)boronic acid

PC32757-1G PC32757-25G PC32757-5G
APOSPC32757-1GEA 622 CZK
APOSPC32757-1G APOSPC32757-25G APOSPC32757-5G
4-Fluoro-2-formylphenylboronic acid
4-Fluoro-2-formylphenylboronic acid
Vzorec: C₇H₆BFO₃
MW: 167,93 g/mol
Bod tání: 123…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697426
CAS číslo: 825644-26-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR