2-Methyl-5-(trifluoromethoxy)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC32734-1G
PC32734-250MG
APOSPC32734-1GEA
2690
CZK
APOSPC32734-1G
APOSPC32734-250MG
2-Methyl-5-(trifluoromethoxy)benzoic acid
2-Methyl-5-(trifluoromethoxy)benzoic acid
Vzorec:
C₉H₇F₃O₃ MW: 220,147 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399742 CAS číslo: 195622-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...