Ethyl perfluorononanoate 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC3253-25G
PC3253-100G
APOSPC3253-25GEA
5200
CZK
APOSPC3253-25G
APOSPC3253-100G
Ethyl perfluorononanoate 97%
Ethyl perfluorononanoate
Vzorec:
C₁₁H₅F₁₇O₂ Bod varu: 84…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039245 CAS číslo: 30377-52-7 EINECS: 250-159-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...