Tisk…

Dimethyl (4-fluorobenzyl)malonate

Dodavatel: Apollo Scientific

PC32416-25G PC32416-5G PC32416-1G
APOSPC32416-25GEA 24600 CZK
APOSPC32416-25G APOSPC32416-5G APOSPC32416-1G
Dimethyl (4-fluorobenzyl)malonate
Dimethyl (4-fluorobenzyl)malonate
Vzorec: C₁₂H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD01934558
CAS číslo: 252938-22-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR