Ethyl heptafluorobutyrate 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Ethyl 2,2,3,3,4,4,4-heptafluorobutanoate, Ethyl perfluorobutyrate
PC3240-25G
PC3240-10G
PC3240-5G
PC3240-100G
APOSPC3240-25GEA
2280
CZK
APOSPC3240-25G
APOSPC3240-10G
APOSPC3240-5G
APOSPC3240-100G
Ethyl heptafluorobutyrate 98%
Ethyl heptafluorobutyrate
Vzorec:
C₆H₅F₇O₂ MW: 242,09 g/mol Bod varu: 95…98 °C Hustota: 1,396 g/cm³ (25 °C) Bod vzplanutí: 14 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000434 CAS číslo: 356-27-4 EINECS: 206-602-1 UN: 3272 ADR: 3,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...