Tisk…

4-Cyano-2-fluorophenylboronic acid pinacol ester 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyano-2-fluorobenzeneboronic acid pinacol ester

PC3223-1G PC3223-250MG
APOSPC3223-1GEA 8240 CZK
APOSPC3223-1G APOSPC3223-250MG
4-Cyano-2-fluorophenylboronic acid pinacol ester 98%
4-Cyano-2-fluorophenylboronic acid pinacol ester
Vzorec: C₁₃H₁₅BFNO₂
MW: 247,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998162
CAS číslo: 1035235-29-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR