Tisk…

4-Cyano-2-fluorophenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

PC3222-1G PC3222-250MG
APOSPC3222-1GEA 4300 CZK
APOSPC3222-1G APOSPC3222-250MG
4-Cyano-2-fluorophenylboronic acid 98%
4-Cyano-2-fluorophenylboronic acid
Vzorec: C₇H₅BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD07644472
CAS číslo: 1150114-77-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR