Tisk…

4-Amino-2-fluorophenylboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PC3221-5G
APOSPC3221-5GEA 2780 CZK
APOSPC3221-5G
4-Amino-2-fluorophenylboronic acid pinacol ester
4-Amino-2-fluorophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₇BFNO₂
MW: 237,08 g/mol
Bod tání: 163…166 °C
Storage Temperature: Freezer
MDL Number: MFCD09951877
CAS číslo: 819057-45-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR