2-[(4-Fluorophenoxy)methyl]oxirane
Dodavatel: Apollo Scientific
Synonyms:
[(4-Fluorophenoxy)methyl]oxirane
PC31966-10G
PC31966-1G
PC31966-5G
APOSPC31966-10GEA
6750
CZK
APOSPC31966-10G
APOSPC31966-1G
APOSPC31966-5G
2-[(4-Fluorophenoxy)methyl]oxirane
2-[(4-Fluorophenoxy)methyl]oxirane
Vzorec:
C₉H₉FO₂ Storage Temperature: Ambient |
CAS číslo:
18123-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...