2-Chlor-1,3-difluor-4-nitrobenzen 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Chloro-2,4-difluoronitrobenzene, 2-Chloro-1,3-difluoro-4-nitrobenzene
PC31739-1G
PC31739-5G
PC31739-10G
APOSPC31739-1GEA
622
CZK
APOSPC31739-1G
APOSPC31739-5G
APOSPC31739-10G
2-Chlor-1,3-difluor-4-nitrobenzen 97%
2-Chlor-1,3-difluor-4-nitrobenzen
Vzorec:
C₆H₂ClF₂NO₂ MW: 193.54 Bod tání: 45…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042201 CAS číslo: 3847-58-3 EINECS: 411-980-8 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...