2-Chloro-6-fluorobenzaldehyde-O-{[(5-chloro-1-methyl-1H-pyrazol-4-yl)amino]carbonyl}oxime
Dodavatel: Apollo Scientific
Synonyms:
5-Chloro-4-{[({[(2-chloro-6-fluorophenyl)methylene]amino}oxy)carbonyl]amino}-1-methyl-1H-pyrazole
PC31702-1G
PC31702-250MG
PC31702-500MG
PC31702-5G
APOSPC31702-1GEA
11180
CZK
APOSPC31702-1G
APOSPC31702-250MG
APOSPC31702-500MG
APOSPC31702-5G
2-Chloro-6-fluorobenzaldehyde-O-{[(5-chloro-1-methyl-1H-pyrazol-4-yl)amino]carbonyl}oxime
2-Chloro-6-fluorobenzaldehyde-O-{[(5-chloro-1-methyl-1H-pyrazol-4-yl)amino]carbonyl}oxime
Vzorec:
C₁₂H₉Cl₂FN₄O₂ MW: 331,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02179999 CAS číslo: 648427-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...