Ethyl chlorodifluoroacetate 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC3170-100G
PC3170-25G
PC3170-5G
APOSPC3170-100GEA
4770
CZK
APOSPC3170-100G
APOSPC3170-25G
APOSPC3170-5G
Ethyl chlorodifluoroacetate 98%
Ethyl chlorodifluoroacetate
Vzorec:
C₄H₅ClF₂O₂ Bod varu: 96…97 °C Hustota: 1,252 g/cm³ (25 °C) Bod vzplanutí: 18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013662 CAS číslo: 383-62-0 EINECS: 206-850-0 UN: 2924 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...