Tisk…

α,3-Dibromo-2-fluorotoluene

Dodavatel: Apollo Scientific
Nebezpečí

PC3168-1G PC3168-5G PC3168-25G
APOSPC3168-1GEA 622 CZK
APOSPC3168-1G APOSPC3168-5G APOSPC3168-25G
α,3-Dibromo-2-fluorotoluene
α,3-Dibromo-2-fluorotoluene
Vzorec: C₇H₅Br₂F
Bod varu: 60 °C/0,1mmHg
Storage Temperature: Ambient
MDL Number: MFCD09842442
CAS číslo: 149947-16-0
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR