Tisk…

5-Methyl-2-(trifluoromethyl)-3-furansulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC31424-1G PC31424-250MG PC31424-500MG PC31424-2.5G PC31424-5G
APOSPC31424-1GEA 5860 CZK
APOSPC31424-1G APOSPC31424-250MG APOSPC31424-500MG APOSPC31424-2.5G APOSPC31424-5G
5-Methyl-2-(trifluoromethyl)-3-furansulphonyl chloride
5-Methyl-2-(trifluoromethyl)-3-furansulphonyl chloride
Vzorec: C₆H₄ClF₃O₃S
Bod varu: 44…46 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD02677699
CAS číslo: 306935-02-4
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR