Dodecafluorosuberic acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC3132T-25G
PC3132T-5G
APOSPC3132T-25GEA
3540
CZK
APOSPC3132T-25G
APOSPC3132T-5G
Dodecafluorosuberic acid 98%
Dodecafluorosuberic acid
Vzorec:
C₈H₂F₁₂O₄ Bod varu: >250 °C Bod tání: 133…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153146 CAS číslo: 678-45-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...