Tisk…

Undecafluoro(2-methyl-3-oxahexanoyl)fluoride 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Perfluoro(2-methyl-3-oxahexanoyl) fluoride

PC3132G-100G PC3132G-25G PC3132G-5G
APOSPC3132G-100GEA 9860 CZK
APOSPC3132G-100G APOSPC3132G-25G APOSPC3132G-5G
Undecafluoro(2-methyl-3-oxahexanoyl)fluoride 97%
Undecafluoro(2-methyl-3-oxahexanoyl)fluoride

HFPO trimer, tetramer, pentamer also available.

Vzorec: C₆F₁₂O₂
MW: 332,05 g/mol
Bod varu: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00054657
CAS číslo: 2062-98-8
EINECS: 218-173-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR