Tisk…

2-[(3-Fluorobenzyl)oxy]-5-nitrobenzaldehyde

Dodavatel: Apollo Scientific

PC31223-1G PC31223-250MG
APOSPC31223-1GEA 2740 CZK
APOSPC31223-1G APOSPC31223-250MG
2-[(3-Fluorobenzyl)oxy]-5-nitrobenzaldehyde
2-[(3-Fluorobenzyl)oxy]-5-nitrobenzaldehyde
Vzorec: C₁₄H₁₀FNO₄
Storage Temperature: Ambient
MDL Number: MFCD00117030
CAS číslo: 219539-49-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR