3,5-Bis(trifluoromethyl)-DL-phenylalanine 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC3114L-1G
PC3114L-500MG
APOSPC3114L-1GEA
1330
CZK
APOSPC3114L-1G
APOSPC3114L-500MG
3,5-Bis(trifluoromethyl)-DL-phenylalanine 98%
3,5-Bis(trifluoromethyl)-DL-phenylalanine
Vzorec:
C₁₁H₉F₆NO₂ MW: 301.19 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00061309 CAS číslo: 237076-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...