Tisk…

(3,5-Bis(trifluoromethyl)phenyl)acetic acid 98%

Dodavatel: Apollo Scientific
Varování

PC3113G-100G PC3113G-1KG PC3113G-25G
APOSPC3113G-100GEA 2480 CZK
APOSPC3113G-100G APOSPC3113G-1KG APOSPC3113G-25G
(3,5-Bis(trifluoromethyl)phenyl)acetic acid 98%
(3,5-Bis(trifluoromethyl)phenyl)acetic acid
Vzorec: (C₆H₃(CF₃)₂)CH₂COOH
MW: 272,15 g/mol
Bod tání: 121…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00009908
CAS číslo: 85068-33-3
EINECS: 285-295-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR