Tisk…

2,4-Bis(trifluoromethyl)benzyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC3105D-25G PC3105D-5G PC3105D-100G
APOSPC3105D-25GEA 13300 CZK
APOSPC3105D-25G APOSPC3105D-5G APOSPC3105D-100G
2,4-Bis(trifluoromethyl)benzyl chloride 98%
2,4-Bis(trifluoromethyl)benzyl chloride
Vzorec: C₉H₅ClF₆
MW: 262,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083537
CAS číslo: 195136-46-0
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR