Tisk…

5-Bromo-2-ethoxybenzotrifluoride 98%

Dodavatel: Apollo Scientific

Synonyms: 1-Bromo-4-ethoxy-3-(trifluoromethyl)benzene, 4-Bromo-1-ethoxy-2-(trifluoromethyl)benzene

PC3098-25G PC3098-5G PC3098-1G
APOSPC3098-25GEA 8170 CZK
APOSPC3098-25G APOSPC3098-5G APOSPC3098-1G
5-Bromo-2-ethoxybenzotrifluoride 98%
5-Bromo-2-ethoxybenzotrifluoride
Vzorec: C₉H₈BrF₃O
MW: 269,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059521
CAS číslo: 914635-58-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR