2,4-Bis(trifluoromethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC3095-1G
PC3095-5G
PC3095-25G
APOSPC3095-1GEA
1410
CZK
APOSPC3095-1G
APOSPC3095-5G
APOSPC3095-25G
2,4-Bis(trifluoromethyl)benzonitrile 98%
2,4-Bis(trifluoromethyl)benzonitrile
Vzorec:
C₉H₃F₆N MW: 239,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00061225 CAS číslo: 177952-38-4 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...