2,6-Bis(trifluoromethyl)benzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC3089-10G
PC3089-1G
PC3089-5G
APOSPC3089-10GEA
0
CZK
APOSPC3089-10G
APOSPC3089-1G
APOSPC3089-5G
2,6-Bis(trifluoromethyl)benzoic acid 98%
2,6-Bis(trifluoromethyl)benzoic acid
Vzorec:
C₉H₄F₆O₂ MW: 258,12 g/mol Bod tání: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000376 CAS číslo: 24821-22-5 EINECS: 246-479-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...