Tisk…

α,α,α,α',α',α'-Hexafluoro-p-xylene 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1,4-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-p-xylene

PC3080-100G PC3080-1KG PC3080-500G
APOSPC3080-100GEA 787 CZK
APOSPC3080-100G APOSPC3080-1KG APOSPC3080-500G
α,α,α,α',α',α'-Hexafluoro-p-xylene 98%
α,α,α,α',α',α'-Hexafluoro-p-xylene
Vzorec: C₈H₄F₆
MW: 214,11 g/mol
Bod varu: 115…117 °C
Bod tání: –1 °C
Hustota: 1,381 g/cm³ (25 °C)
Bod vzplanutí: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00000402
CAS číslo: 433-19-2
EINECS: 207-086-0
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR