2,4-Bis(trifluoromethyl)benzamide 97%
Dodavatel: Apollo Scientific
PC3064W-5G
PC3064W-10G
PC3064W-1G
APOSPC3064W-5GEA
3410
CZK
APOSPC3064W-5G
APOSPC3064W-10G
APOSPC3064W-1G
2,4-Bis(trifluoromethyl)benzamide 97%
2,4-Bis(trifluoromethyl)benzamide
Vzorec:
C₉H₅F₆NO Storage Temperature: Ambient |
MDL Number:
MFCD00083536 CAS číslo: 53130-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...