Tisk…

5-Bromo-2-fluoroisonicotinic acid

Dodavatel: Apollo Scientific

PC305015-1G PC305015-250MG
APOSPC305015-1GEA 14290 CZK
APOSPC305015-1G APOSPC305015-250MG
5-Bromo-2-fluoroisonicotinic acid
5-Bromo-2-fluoroisonicotinic acid
Vzorec: C₆H₃BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185803
CAS číslo: 1214377-40-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR