Tisk…

3-(Trifluoromethyl)azetidin-3-ol hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Trifluoromethyl-3-azetidinol hydrochloride

PC305004-1G PC305004-250MG
APOSPC305004-1GEA 10950 CZK
APOSPC305004-1G APOSPC305004-250MG
3-(Trifluoromethyl)azetidin-3-ol hydrochloride 98%
3-(Trifluoromethyl)azetidin-3-ol hydrochloride
Vzorec: C₄H₇ClF₃NO
MW: 177,551 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13248564
CAS číslo: 848192-96-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR