Tisk…

Sangerova reagencie 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene

PC3040-100G PC3040-1KG PC3040-500G
APOSPC3040-100GEA 910 CZK
APOSPC3040-100G APOSPC3040-1KG APOSPC3040-500G
Sangerova reagencie 99%
Sangerova reagencie

Nucleophiles can displace the activated fluorine atom.Reagent for labelling of terminal amino acid groups.

Vzorec: C₆H₃FN₂O₄
MW: 186,1 g/mol
Bod tání: 28…30 °C
Hustota: 1,482 g/cm³ (25 °C)
Bod vzplanutí: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007056
CAS číslo: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR