4,6-Difluoro-m-toluic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
2,4-Difluoro-5-methylbenzoic acid
PC303306-5G
PC303306-1G
PC303306-25G
APOSPC303306-5GEA
2570
CZK
APOSPC303306-5G
APOSPC303306-1G
APOSPC303306-25G
4,6-Difluoro-m-toluic acid 97%
4,6-Difluoro-m-toluic acid
Vzorec:
C₈H₆F₂O₂ MW: 172.13 g/mol Bod tání: 159…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD09835285 CAS číslo: 367954-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...