Tisk…

3,4-Dichloro-6-fluoroaniline 97%

Dodavatel: Apollo Scientific

PC303284-1G PC303284-5G
APOSPC303284-1GEA 1910 CZK
APOSPC303284-1G APOSPC303284-5G
3,4-Dichloro-6-fluoroaniline 97%
3,4-Dichloro-6-fluoroaniline
Vzorec: C₆H₄Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD19707630
CAS číslo: 2729-36-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR