3-Bromo-2-methyl-5-(pentafluorothio)aniline 97%
Dodavatel: Apollo Scientific
PC303147-1G
PC303147-5G
APOSPC303147-1GEA
9920
CZK
APOSPC303147-1G
APOSPC303147-5G
3-Bromo-2-methyl-5-(pentafluorothio)aniline 97%
3-Bromo-2-methyl-5-(pentafluorothio)aniline
Vzorec:
C₇H₇BrF₅NS MW: 312,10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201077 CAS číslo: 1373920-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...