Tisk…

5-Methyl-2-(trifluoromethoxy)aniline 98%

Dodavatel: Apollo Scientific

Synonyms: 6-(Trifluoromethoxy)-m-toluidine, 5-Methyl-2-(trifluoromethoxy)benzenamine, Benzenamine, 5-methyl-2-(trifluoromethoxy)-

PC303143-1G PC303143-5G
APOSPC303143-1GEA 9000 CZK
APOSPC303143-1G APOSPC303143-5G
5-Methyl-2-(trifluoromethoxy)aniline 98%
5-Methyl-2-(trifluoromethoxy)aniline
Vzorec: C₈H₈F₃NO
MW: 191,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399696
CAS číslo: 151276-15-2
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR