4-Methoxy-3-methyl-5-(trifluoromethyl)benzamide 97%
Dodavatel: Apollo Scientific
Synonyms:
Benzamide, 4-methoxy-3-methyl-5-(trifluoromethyl)-
PC303116-5G
PC303116-1G
APOSPC303116-5GEA
9000
CZK
APOSPC303116-5G
APOSPC303116-1G
4-Methoxy-3-methyl-5-(trifluoromethyl)benzamide 97%
4-Methoxy-3-methyl-5-(trifluoromethyl)benzamide
Vzorec:
C₁₀H₁₀F₃NO₂ MW: 233,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22682823 CAS číslo: 1431329-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...