4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline 97%
Dodavatel: Apollo Scientific
PC303109-10G
PC303109-5G
PC303109-1G
APOSPC303109-10GEA
11540
CZK
APOSPC303109-10G
APOSPC303109-5G
APOSPC303109-1G
4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline 97%
4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline
Vzorec:
C₁₃H₉F₄NO MW: 271,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08687913 CAS číslo: 946784-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...