3-Fluoro-4-(4-methoxyphenoxy)phenylamine
Dodavatel: Apollo Scientific
Synonyms:
3-Fluoro-4-(4-methoxyphenoxy)aniline
PC303107-10G
PC303107-5G
APOSPC303107-10GEA
11540
CZK
APOSPC303107-10G
APOSPC303107-5G
3-Fluoro-4-(4-methoxyphenoxy)phenylamine
3-Fluoro-4-(4-methoxyphenoxy)phenylamine
Vzorec:
C₁₃H₁₂FNO₂ MW: 233,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08687845 CAS číslo: 87294-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...