Tisk…

4-Chloro-3-(trifluoromethoxy)benzonitrile 98%

Dodavatel: Apollo Scientific

PC303080-5G PC303080-1G
APOSPC303080-5GEA 3310 CZK
APOSPC303080-5G APOSPC303080-1G
4-Chloro-3-(trifluoromethoxy)benzonitrile 98%
4-Chloro-3-(trifluoromethoxy)benzonitrile
Vzorec: C₈H₃ClF₃NO
MW: 221,56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06660261
CAS číslo: 886501-50-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR