4-Chloro-2-(trifluoromethoxy)benzoic acid 98%
Dodavatel: Apollo Scientific
PC303076-10G
PC303076-5G
PC303076-1G
APOSPC303076-10GEA
10250
CZK
APOSPC303076-10G
APOSPC303076-5G
APOSPC303076-1G
4-Chloro-2-(trifluoromethoxy)benzoic acid 98%
4-Chloro-2-(trifluoromethoxy)benzoic acid
Vzorec:
C₈H₄ClF₃O₃ MW: 240,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18393801 CAS číslo: 1261605-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...