Tisk…

3-Chloro-4-(trifluoromethyl)benzylamine 98%

Dodavatel: Apollo Scientific

PC303057-1G PC303057-250MG
APOSPC303057-1GEA 7440 CZK
APOSPC303057-1G APOSPC303057-250MG
3-Chloro-4-(trifluoromethyl)benzylamine 98%
3-Chloro-4-(trifluoromethyl)benzylamine
Vzorec: C₈H₇ClF₃N
MW: 209,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972759
CAS číslo: 361393-93-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR