Tisk…

2-Methyl-5-(trifluoromethoxy)benzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC303049-10G PC303049-5G PC303049-1G
APOSPC303049-10GEA 15940 CZK
APOSPC303049-10G APOSPC303049-5G APOSPC303049-1G
2-Methyl-5-(trifluoromethoxy)benzyl bromide
2-Methyl-5-(trifluoromethoxy)benzyl bromide
Vzorec: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD18399774
CAS číslo: 1261573-60-7
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR